Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1269200

USP

Famotidine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Famotidine, N′-(Aminosulfonyl)-3-([2-(diaminomethyleneamino)-4-thiazolyl]methylthio)propanamidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15N7O2S3
CAS Number:
Molecular Weight:
337.45
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

USP

vapor pressure

<0.0000001 kPa ( 25 °C)

API family

famotidine

form

powder

packaging

pkg of 125 mg

manufacturer/tradename

USP

storage condition

protect from light

color

white to pale yellow-white

mp

325.4-327.2 °F (163—164°C)

solubility

DMF: freely soluble
acetone: practically insoluble
chloroform: practically insoluble
ethanol: practically insoluble
ether: practically insoluble
ethyl acetate: practically insoluble
glacial acetic acid: freely soluble
methanol: slightly soluble
water: very slightly soluble

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N\C(N)=N\c1nc(CSCCC(=N)NS(N)(=O)=O)cs1

InChI

1S/C8H15N7O2S3/c9-6(15-20(12,16)17)1-2-18-3-5-4-19-8(13-5)14-7(10)11/h4H,1-3H2,(H2,9,15)(H2,12,16,17)(H4,10,11,13,14)

InChI key

XUFQPHANEAPEMJ-UHFFFAOYSA-N

Gene Information

human ... HRH2(3274)

Looking for similar products? Visit Product Comparison Guide

General description

Famotidine is a hydrophilic, cationic, histamine H2 receptor antagonist drug that effectively inhibits gastric acid secretion in humans.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Famotidine can be used as a pharmaceutical primary standard for the determination of the analyte in pharmaceutical formulations by various analytical techniques.(10)

Biochem/physiol Actions

H2 histamine receptor antagonist; anti-ulcer agent

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Simultaneous determination of famotidine and flurbiprofen by high performance liquid chromatography
Hanif M, et al.
Tropical Journal of Pharmaceutical Research, 15(3), 605-611 (2016)
Determination of cimetidine, famotidine, and ranitidine hydrochloride in the presence of their sulfoxide derivatives in pure and dosage forms by high-performance thin-layer chromatography and scanning densitometry
Kelani KM, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 85(5), 1015-1020 (2002)
Stability indicating method for famotidine in pharmaceuticals using porous graphitic carbon column
Helali N and Monser L
Journal of Separation Science, 31(2) (2008)
Capillary zone electrophoresis method for the determination of famotidine and related impurities in pharmaceuticals
Helali N, et al.
Talanta, 74(4) (2008)
Famotidine
Pharmacopeia, US
United States Pharmacopeia, 39(1)(1-2), 1815-1815 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service