Skip to Content
MilliporeSigma

1268819

USP

Etoposide Phosphate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(5R,5aR,8aR,9S)-9-[4,6-O-(R)-Ethylidene-D-glucopyranosyloxy]-5,8,8a,9-tetrahydro-5-[3,5-dimethoxy-4-(phosphonooxy)phenyl]furo[3’,4’:6,7]naphtho[2,3-d]-1,3-dioxol-6(5aH)-one)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H33O16P
CAS Number:
Molecular Weight:
668.54
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

etoposide

form

solid

manufacturer/tradename

USP

SMILES string

[P](=O)(Oc1c(cc(cc1OC)[C@H]2[C@@H]3[C@@H]([C@@H](c6c2cc7c(c6)OCO7)O[C@@H]4O[C@H]5[C@@H](O[C@@H](OC5)C)[C@@H]([C@H]4O)O)COC3=O)OC)(O)O

InChI

1S/C29H33O16P/c1-11-38-9-20-27(42-11)23(30)24(31)29(43-20)44-25-14-7-17-16(40-10-41-17)6-13(14)21(22-15(25)8-39-28(22)32)12-4-18(36-2)26(19(5-12)37-3)45-46(33,34)35/h4-7,11,15,20-25,27,29-31H,8-10H2,1-3H3,(H2,33,34,35)/t11-,15+,20-,21-,22+,23-,24-,25-,27-,29+/m1/s1

InChI key

LIQODXNTTZAGID-OCBXBXKTSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Etoposide Phosphate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Etoposide Phosphate for Injection


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library