Skip to Content
MilliporeSigma

1166535

USP

6,7-Dehydronorethindone acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3-Oxo-19-nor-17α-pregn-4,6-dien-20-yn-17-yl acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H26O3
CAS Number:
Molecular Weight:
338.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

norethindrone

packaging

pkg of 20 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O([C@@]1([C@@]2([C@H]([C@H]3[C@@H]([C@H]4CCC(=O)C=C4C=C3)CC2)CC1)C)C#C)C(=O)C

InChI

1S/C22H26O3/c1-4-22(25-14(2)23)12-10-20-19-7-5-15-13-16(24)6-8-17(15)18(19)9-11-21(20,22)3/h1,5,7,13,17-20H,6,8-12H2,2-3H3/t17-,18+,19+,20-,21-,22-/m0/s1

InChI key

DQSVONCNGQQILU-ZCPXKWAGSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

6,7-Dehydronorethindone acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library