Skip to Content
MilliporeSigma

Skip To

1129007

USP

Chlortetracycline hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

7-Chlorotetracycline hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C22H23ClN2O8 · HCl
CAS Number:
Molecular Weight:
515.34
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3858364
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

chlortetracycline

form

solid

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2CC3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)ccc(Cl)c4[C@@]3(C)O

InChI

1S/C22H23ClN2O8.ClH/c1-21(32)7-6-8-15(25(2)3)17(28)13(20(24)31)19(30)22(8,33)18(29)11(7)16(27)12-10(26)5-4-9(23)14(12)21;/h4-5,7-8,15,26,28-29,32-33H,6H2,1-3H3,(H2,24,31);1H/t7?,8-,15-,21-,22-;/m0./s1

InChI key

CBHYYLPALVVVEY-LYNLVHCPSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Chlortetracycline hydrochloride is a first-generation tetracycline antibiotic with activity against a wide spectrum of bacteria. It functions by preventing aminoacyl-tRNA attachment to the bacterial ribosome. The USP standard aids in identity verification and quantitative assay testing.

The USP biologics antibiotics category includes a wide range of antimicrobial agents that are essential in treating bacterial infections. These antibiotics are derived from various natural sources or synthesized to combat specific pathogens effectively. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of antibiotic therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Chlortetracycline hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Chlortetracycline Hydrochloride Ointment
  • Chlortetracycline Hydrochloride Tablets
  • Chlortetracycline Hydrochloride Soluble Powder
  • Chlortetracycline Hydrochloride Ophthalmic Ointment
  • Chlortetracycline and Sulfamethazine Bisulfates Soluble Powder
  • Chlortetracycline Bisulfate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of the USP Biologics program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopeia.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1520C2000000BP783
description

United States Pharmacopeia (USP) Reference Standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

European Pharmacopoeia (EP) Reference Standard

description

British Pharmacopoeia (BP) Reference Standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

BP

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

form

solid

form

-

form

-

form

solid

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

format

neat

format

neat

format

neat

API family

chlortetracycline

API family

chlortetracycline

API family

chlortetracycline

API family

chlortetracycline


Still not finding the right product?

Explore all of our products under Chlortetracycline hydrochloride


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters