Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
chlorhexidine
manufacturer/tradename
USP
mp
134-136 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC(=N)Nc2ccc(Cl)cc2)cc1
InChI
1S/C22H30Cl2N10/c23-15-5-9-17(10-6-15)31-21(27)33-19(25)29-13-3-1-2-4-14-30-20(26)34-22(28)32-18-11-7-16(24)8-12-18/h5-12H,1-4,13-14H2,(H5,25,27,29,31,33)(H5,26,28,30,32,34)
InChI key
GHXZTYHSJHQHIJ-UHFFFAOYSA-N
General description
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description United States Pharmacopeia (USP) Reference Standard | description European Pharmacopoeia (EP) Reference Standard | description Pharmaceutical Secondary Standard; Certified Reference Material | description ≥99.5% |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade - |
| manufacturer/tradename USP | manufacturer/tradename EDQM | manufacturer/tradename - | manufacturer/tradename - |
| format neat | format neat | format neat | format - |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) - |
| storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C | storage temp. - |
| mp 134-136 °C (lit.) | mp 134-136 °C (lit.) | mp 134-136 °C (lit.) | mp 134-136 °C (lit.) |
Still not finding the right product?
Explore all of our products under Chlorhexidine
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


