Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
(C6H5O8)2 · Ca3 · 4H2O
CAS Number:
Molecular Weight:
602.49
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
grade
pharmaceutical primary standard
agency
USP
API family
calcium (-)-hydroxycitrate
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
15-25°C
SMILES string
[Ca+2].[Ca+2].[Ca+2].[O-]C(=O)[C@@](O)([C@H](O)C(=O)[O-])CC(=O)[O-].[O-]C(=O)[C@@](O)([C@H](O)C(=O)[O-])CC(=O)[O-]
InChI
1S/2C6H8O8.3Ca/c2*7-2(8)1-6(14,5(12)13)3(9)4(10)11;;;/h2*3,9,14H,1H2,(H,7,8)(H,10,11)(H,12,13);;;/q;;3*+2/p-6/t2*3-,6+;;;/m11.../s1
InChI key
RVEPHTVUPANNSH-IFDHGPESSA-H
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Calcium (−)-hydroxycitrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
- Powdered Garcinia Hydroxycitrate Extract
- Powdered Garcinia cambogia
- Garcinia cambogia
- Powdered Garcinia indica
- Garcinia indica
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.