Skip to Content
MilliporeSigma

1075815

USP

Bisoprolol Related Compound F

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Isopropoxyethyl-4-[2-hydroxy-3-(isopropylamino)propoxy]benzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H29NO5
CAS Number:
Molecular Weight:
339.43
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

bisoprolol

form

solid

manufacturer/tradename

USP

SMILES string

N(C(C)C)CC(O)COc1ccc(cc1)C(=O)OCCOC(C)C

InChI

1S/C18H29NO5/c1-13(2)19-11-16(20)12-24-17-7-5-15(6-8-17)18(21)23-10-9-22-14(3)4/h5-8,13-14,16,19-20H,9-12H2,1-4H3

InChI key

DUPZZHDDPRYPMB-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bisoprolol Related Compound F USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library