Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1022808

USP

2-Amino-5-chlorobenzophenone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Chloro-2-benzoylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(Cl)COC6H5
CAS Number:
Molecular Weight:
231.68
Beilstein:
475640
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

description

API family: alprazolam

API family

chlordiazepoxide , alprazolam, oxazepam; prazepam

manufacturer/tradename

USP

mp

96-98 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ccc(Cl)cc1C(=O)c2ccccc2

InChI

1S/C13H10ClNO/c14-10-6-7-12(15)11(8-10)13(16)9-4-2-1-3-5-9/h1-8H,15H2

InChI key

ZUWXHHBROGLWNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Amino-5-chlorobenzophenone is a organic nonlinear optical material.

Application

It was used in the determination of chlordiazepoxide and mebeverine HCl using HPLC method.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

388.4 °F - closed cup

Flash Point(C)

198 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on organic single crystal: 2-amino-5-chlorobenzophenone (2A-5CB).
Babu, R. Ramesh, et al.
Journal of Crystal Growth, 265.1, 290-295 (2004)
Simultaneous HPLC Determination of Chlordiazepoxide and Mebeverine HCl in the Presence of Their Degradation Products and Impurities.
El-Shaheny, Rania N., and Fathalla F. Belal.
Journal of Chemistry, 2015, 1-9 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service