1012495
USP
beta-Alanine
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
β-Alanine, β-Ala, 3-Aminopropionic acid
About This Item
grade
pharmaceutical primary standard
API family
beta-alanine
manufacturer/tradename
USP
mp
202 °C (dec.) (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
NCCC(O)=O
InChI
1S/C3H7NO2/c4-2-1-3(5)6/h1-2,4H2,(H,5,6)
InChI key
UCMIRNVEIXFBKS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Analysis Note
Other Notes
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service