Skip to Content

1012101

USP

Adenine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

6-Aminopurine, Vitamin B4

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H5N5
CAS Number:
Molecular Weight:
135.13
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5777
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

adenine

form

powder

manufacturer/tradename

USP

mp

>360 °C (lit.)

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ncnc2[nH]cnc12

InChI

1S/C5H5N5/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H3,6,7,8,9,10)

InChI key

GFFGJBXGBJISGV-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Adenine is a purine nucleobase that is a fundamental component of nucleic acids, including DNA and RNA. In the context of oligonucleotide synthesis, adenine is incorporated into synthetic DNA or RNA sequences. The USP Adenine standard ensures the purity and consistency of adenine used in the synthesis of oligonucleotides for research, therapeutic, and diagnostic purposes.

The USP biologics oligonucleotides category includes a variety of synthetic nucleic acid sequences that are essential in research, diagnostics, and therapeutic applications. These oligonucleotides play crucial roles in molecular biology, including gene expression regulation, antisense therapy, and RNA interference. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of oligonucleotide products throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Adenine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Anticoagulant Citrate Phosphate Dextrose Adenine Solution
  • Adenosine
  • S-Adenosyl-l-methionine 1
  • 4-Butanedisulfonate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of USP Biologics Program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.


Still not finding the right product?

Explore all of our products under Adenine


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library