Skip to Content
MilliporeSigma

1001502

USP

Acepromazine maleate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Acetopromazine maleate salt, 2-Acetyl-10-(3-dimethylaminopropyl)phenothiazine maleate salt, Acepromazine maleate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H22N2OS · C4H4O4
CAS Number:
Molecular Weight:
442.53
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

acepromazine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)\C=C/C(O)=O.CN(C)CCCN1c2ccccc2Sc3ccc(cc13)C(C)=O

InChI

1S/C19H22N2OS.C4H4O4/c1-14(22)15-9-10-19-17(13-15)21(12-6-11-20(2)3)16-7-4-5-8-18(16)23-19;5-3(6)1-2-4(7)8/h4-5,7-10,13H,6,11-12H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

FQRHOOHLUYHMGG-BTJKTKAUSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acepromazine maleate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Acepromazine Maleate Injection
  • Acepromazine Maleate Tablets

Biochem/physiol Actions

Acepromazine is a phenothiazine antipsychotic comonly used as a veterinary drug (horses, dogs and cats). The compound is an analogue antipsychotic drug chlorpromazine (#C8138). The drug is thought to block receptors of dopamine in the brain. Recently it was discover that the compound inhibits ABCG2 transported protein, which overexpression in tumors is associated with drug resistance.
Atypical neuroleptic.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Acepromazine maleate


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Elizabeth A Nunamaker et al.
Journal of the American Association for Laboratory Animal Science : JAALAS, 53(5), 494-501 (2014-09-26)
The goal of the current study was to compare the efficacy, adverse effects, and plasma buprenorphine concentrations of sustained-release buprenorphine (SRB) and buprenorphine after subcutaneous administration in dogs undergoing ovariohysterectomy. In a prospective, randomized, blinded design, 20 healthy adult female
Samer M Jaber et al.
Journal of the American Association for Laboratory Animal Science : JAALAS, 53(6), 684-691 (2015-02-05)
Extending a surgical plane of anesthesia in mice by using injectable anesthetics typically is accomplished by repeat-bolus dosing. We compared the safety and efficacy of redosing protocols administered either during an anesthetic surgical plane (maintaining a continuous surgical plane, CSP)
Cathryn M Kolka et al.
Metabolism: clinical and experimental, 64(2), 330-337 (2014-12-04)
Insulin injected directly into skeletal muscle diffuses rapidly through the interstitial space to cause glucose uptake, but this is blocked in insulin resistance. As glucotoxicity is associated with endothelial dysfunction, the observed hyperglycemia in diet-induced obese dogs may inhibit insulin



Global Trade Item Number

SKUGTIN
1001502-250MG04061833859018