Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z777303

Unix SEC-200 (1.8 µm) UHPLC Columns

L x I.D. 15 cm × 4.6 mm, neutral, hydrophilic film bonded to silica phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
SB.52

product name

Unix SEC-200 UHPLC Column, L × I.D. 15 cm × 4.6 mm, 1.8 μm particle size

material

stainless steel column

Agency

suitable for USP L59

description

protein molecular weight range - 5,000 - 1,250,000 Da

product line

Unix

manufacturer/tradename

Unix

packaging

1 ea of

parameter

310 bar pressure (4500 psi)
~80 °C temperature

technique(s)

UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

spherical silica
fully porous particle

matrix active group

neutral, hydrophilic film bonded to silica phase

particle size

1.8 μm

pore size

200 Å average pore size

pH range

2.0-8.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Utilizing proprietary surface technologies, Unix SEC-200 phase is made of uniform, hydrophilic, and neutral nanometer thick films chemically bonded to high purity and mechanically stabilized silica with the particle size of 1.8 μm. The combination of small particle size and large pore volume of Unix SEC-300 renders the highest separation effciency and resolution of analytes. The well controlled surface chemisty results in excellent lot-to-lot reproducibility. The unique bonding chemistry, coupled with the maximized bonding density, allows Unix SEC-200 to provide high stability and negligible non-specific interactions. Typical applications for the Unix SEC-200 columns include separation and analysis of biological molecules and water-soluble polymers.

Legal Information

Unix is a trademark of Sepax Technologies

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service