Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z777213

Proteomix® SAX-NP1.7 (1.7 µm) HPLC Columns

L × I.D. 3 cm × 4.6 mm, stainless steel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Proteomix® SAX-NP1.7 Strong Anion Exchange Column, L × I.D. 3 cm × 4.6 mm, 1.7 μm particle size, stainless steel

material

stainless steel

product line

Proteomix®

manufacturer/tradename

Sepax 403NP2-4603

packaging

1 ea of

parameter

69 Mpa max. pressure (10000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

3 cm × 4.6 mm

matrix

spherical PS-DVB
polymeric

matrix active group

ammonium ion, quaternary phase

particle size

1.7 μm

pore size

non-porous

operating pH range

2.0-12.0

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Proteomix columns are packed with poly(styrene divinylbenzene) particles that are shielded by a hydrophilic polymer layer that contains a network of weak (WAX) or strong (SAX) anion exchange groups. Use Proteomix anion-exchange columns for the analyis or small scale purification of negatively charged biopolymers, such as glycoprotein isoforms, pegylated proteins and oligonucleotides.

Proteomix User Manual

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service