Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z777134

Proteomix® SCX-NP10 (10 µm) HPLC Columns

L × I.D. 1 cm × 4 mm, stainless steel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Proteomix® SCX-NP10 Strong Cation Exchange Guard Cartridge and Holder, L × I.D. 1 cm × 4 mm, 10 μm particle size, stainless steel

material

stainless steel

Agency

suitable for USP L22

product line

Proteomix®

manufacturer/tradename

Sepax 401NP10-4001C

packaging

1 ea of

parameter

27.5 Mpa max. pressure (4000 psi)
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

1 cm × 4 mm

matrix

spherical PS-DVB
polymeric

matrix active group

sulfonic acid phase

particle size

10 μm

pore size

non-porous

operating pH range

2.0-12.0

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

The bonded phase in SRT-C SEC columns is more polar than the hydrophilic phase used in SRT columns, making SRT-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies. SRT-C columns share with SRT columns the same 5 micron silica particle and the same wide range of pore sizes so you can select the pore size that will allow full access for your compound of interest.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service