Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z226165

Nucleosil® C18 HPLC Column

3 μm particle size, L × I.D. 10 cm × 4.6 mm

Synonym(s):

Nucleosil C18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Agency

suitable for USP L1

Quality Level

product line

Nucleosil®

feature

endcapped

manufacturer/tradename

Nucleosil®

extent of labeling

15% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

10 cm × 4.6 mm

surface area

350 m2/g

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

3 μm

pore size

100 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Application



  • Purification and Product Characterization of Lipoxygenase from Opium Poppy Cultures (Papaver somniferum L.).: This research focused on the purification and characterization of lipoxygenase from opium poppy cultures using HPLC with a Nucleosil® C18 column. The study provided detailed insights into the enzyme′s properties and potential applications in pharmaceutical industries (Holková et al., 2019).


  • A Rapid Stability-Indicating RP-HPLC Method for the Determination of Betaxolol Hydrochloride in Pharmaceutical Tablets.: The study developed a rapid, stability-indicating reverse-phase HPLC method for determining betaxolol hydrochloride in tablets using a Nucleosil® C18 column. The method was validated for its accuracy, precision, and robustness (Auvity et al., 2013).


  • Conversion L-tryptophan to melatonin in the gastrointestinal tract: the new high performance liquid chromatography method enabling simultaneous determination of six metabolites of L-tryptophan by native fluorescence and UV-VIS detection.: This study developed a novel HPLC method with a Nucleosil® C18 column for the simultaneous determination of six metabolites of L-tryptophan, providing a comprehensive approach to studying melatonin synthesis in the gastrointestinal tract (Zagajewski et al., 2012).


  • Development and validation of an HPLC-method for determination of free and bound phenolic acids in cereals after solid-phase extraction.: The research presented a validated HPLC method using a Nucleosil® C18 column for determining phenolic acids in cereals. The method included solid-phase extraction, ensuring high sensitivity and specificity for both free and bound phenolic acids (Irakli et al., 2012).


Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service