Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0002338

Supelco

Crotamiton for peak identification

CRS, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-Ethyl-o-crotonotoluidide, N-Ethyl-N-(2-methylphenyl)-2-butenamide, Crotamiton

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCON(C2H5)C6H4CH3
CAS Number:
Molecular Weight:
203.28
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

description

predominantly trans

API family

crotamiton

manufacturer/tradename

EDQM

refractive index

n20/D 1.54 (lit.)

bp

153-155 °C/13 mmHg (lit.)

density

0.987 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

storage temp.

2-8°C

SMILES string

CCN(C(=O)\C=C\C)c1ccccc1C

InChI

1S/C13H17NO/c1-4-8-13(15)14(5-2)12-10-7-6-9-11(12)3/h4,6-10H,5H2,1-3H3/b8-4+

InChI key

DNTGGZPQPQTDQF-XBXARRHUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Crotamiton for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

Unit quantity: 1mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service