Skip to Content
MilliporeSigma

SPRF6

Smoke Point Reference Fuel Blend

Fuel Blend 6 for 35.4 mm (5/95%v/v), certified reference material

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd



Still not finding the right product?

Explore all of our products under Smoke Point Reference Fuel Blend


Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class

3 - Flammable liquids

Flash Point (°F)

10.4 °F - closed cup

Flash Point (°C)

-12 °C - closed cup

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library