Skip to Content
MilliporeSigma
All Photos(1)

Documents

RAD166

radiello Cartridge Adsorbents

for sampling HF, NO2, and SO2, matrix microporous PE impregnated with wet TEA, pk of 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

Agency

EPA IP-5C
NIOSH 6700
NIOSH 7906

packaging

pk of 20

manufacturer/tradename

Radiello

technique(s)

passive air sampling: suitable

matrix

microporous PE impregnated with wet TEA

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling HF, NO2, and SO2

Looking for similar products? Visit Product Comparison Guide

General description

Several different Radiello cartridge adsorbents are available. Each of which are specific for sampling different classes of compounds. The dimensions for each cartridge are 60 mm L x 4.8 or 5.8 mm D. Each cartridge is designed for one time use with the exception of Radiello thermal desorption (TD) cartridge adsorbents.

Each cartridge arrives in a sealed glass or plastic tube wrapped in a transparent thermally sealed polyethylene (PE) bag. The same sealed tube can be used to store the cartridge after sampling prior to desorption/analysis. A sufficient number of adhesive barcode labels are included with every pack of cartridges for easy tracking of sampling date and time.

Note: To conduct air sampling using the Radiello system, Radiello diffusive bodies and triangular support plates must be ordered (separately) in addition to cartridge adsorbents. Radiello Starter Kits (RADxxxS) include all the necessary parts to complete a sampling.

Legal Information

radiello is a trademark of Institi Clinici Scientifici Maugeri

Disclaimer

*Note: This product is not officially specified in the method, it may be suitable for use for the compound(s) listed in the method*

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

354.2 °F - closed cup

Flash Point(C)

179 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
Mayenite based supports for atmospheric NOx sampling
Cucciniello, R., et al.
Atmospheric Environment, 79, 666-671 (2013)
Monitoring of NO2 in the ambient air with passive samplers before and after a road reconstruction event
Stranger, M., et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 90 (2), 93-98 (2008)
A Piechocki-Minguy et al.
The Science of the total environment, 366(1), 55-64 (2005-09-13)
Personal NO(2) exposure measurements were achieved during two campaigns in a large northern France city. These campaigns were following an innovating approach based on sequential exposure measurements by diffusive samplers distinguishing four categories of microenvironments ("home", "other indoor places", "transport"

Articles

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Learn the how the diffusive air sampler works to see if it’s the right fit for you.

Protocols

Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.

Related Content

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service