Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR9025

Supelco

Methyl 4-aminobenzoate

reference material

Synonym(s):

Benzocaine impurity H (PhEur)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2CH3
CAS Number:
Molecular Weight:
151.16
Beilstein:
775913
MDL number:
UNSPSC Code:
41116107

grade

reference material

API family

benzocaine

Assay

≥97.0% (GC)

form

crystals

shelf life

limited shelf life, expiry date on the label

mp

110-111 °C (lit.)

application(s)

pharmaceutical small molecule

SMILES string

COC(=O)c1ccc(N)cc1

InChI

1S/C8H9NO2/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5H,9H2,1H3

InChI key

LZXXNPOYQCLXRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service