Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL89794

Sinomenine hydrochloride

phyproof® Reference Substance

Synonym(s):

(9a,13a,14a)-7,8-Didehydro-4-hydroxy-3,7-dimethoxy-17-methyl-morphinan-6-one hydrochloride, Cucoline hydrochloride, Kukolin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO4 · HCl
CAS Number:
Molecular Weight:
365.85
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

OC1=C(OC)C=CC(C2)=C1[C@]3(CC(C(OC)=C4)=O)[C@@]4([H])[C@H]2N(C)CC3.Cl

InChI

1S/C19H23NO4.ClH/c1-20-7-6-19-10-14(21)16(24-3)9-12(19)13(20)8-11-4-5-15(23-2)18(22)17(11)19;/h4-5,9,12-13,22H,6-8,10H2,1-3H3;1H/t12-,13+,19-;/m1./s1

InChI key

YMEVIMJAUHZFMW-VUIDNZEBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Sinomenine is a herbal medicine used to treat inflammatory diseases like rheumatoid arthritis. Sinomenine had anti-inflammatory effects in several animal models of inflammation, and inhibits mast cell degranulation and histamine release. Additionally, Sinomenine potentiates renewal of human embryonic stem cells.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service