Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL89734

16-O-Methylcafestol

phyproof® Reference Substance

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

CO[C@]1(CO)C[C@@]23CCC4c5ccoc5CC[C@@]4(C)C2CCC1C3

InChI

1S/C21H30O3/c1-19-8-6-17-15(7-10-24-17)16(19)5-9-20-11-14(3-4-18(19)20)21(12-20,13-22)23-2/h7,10,14,16,18,22H,3-6,8-9,11-13H2,1-2H3/t14-,16-,18+,19-,20+,21+/m1/s1

InChI key

BDVVNPOGDNWUOI-GVOJMRIRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

  • 16-O-Methylcafestol biochemical research applications: A study utilized benchtop NMR spectroscopy to quantify 16-O-methylcafestol in lipophilic extracts of ground roast coffee, showcasing its potential as a biochemical assay reagent and highlighting its importance in coffee chemistry research (Gunning et al., 2023).
  • 16-O-Methylcafestol diterpene compound studies: Research focused on the quantitative NMR methodology for the authentication of roasted coffee has highlighted the role of 16-O-methylcafestol as a significant compound in the verification and blend prediction of coffee products, demonstrating its application in food chemistry and bioactive compound analysis (Burton et al., 2020).
  • 16-O-Methylcafestol in vitro biological activity analysis: An ultra-high-performance liquid chromatography-tandem mass spectrometry method was optimized and validated for analyzing 16-O-methylcafestol, emphasizing its pharmacological properties and importance in coffee bean derivative research and potential therapeutic applications (Guercia et al., 2020).

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service