Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHL89172

(+)-Catechin

phyproof® Reference Substance

Synonym(s):

(+)-Cyanidanol, D-Catechin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14O6
CAS Number:
Molecular Weight:
290.27
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3

InChI

1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15+/m0/s1

InChI key

PFTAWBLQPZVEMU-DZGCQCFKSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Improving structural and functional properties of starch-catechin-based green nanofiber mats for active food packaging by electrospinning and crosslinking techniques: This study highlights the application of (+)-Catechin in the development of biodegradable food packaging materials, utilizing its antioxidant properties to enhance the shelf life and safety of food products (Lv et al., 2024).

  • Effect of geographical origin on the chemical characteristics of Q. canariensis acorns: profiling fatty acids, tocopherols, and phenolic composition: Research on (+)-Catechin′s impact on the nutritional profile of acorns from various locations, providing valuable data for food science and agro-ecological studies (Fkiri et al., 2024).

  • Discrimination and prediction of Qingzhuan tea storage year using quantitative chemical profile combined with multivariate analysis: This study utilizes (+)-Catechin to determine the age of Qingzhuan tea through advanced chemical profiling techniques, demonstrating its importance in the quality control and authentication of traditional teas (Li et al., 2024).

Other Notes

This compound is commonly found in plants of the genus: cinnamomum crataegus eucalyptus hypericum primula

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service