Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL85751

Isorhoifolin

phyproof® Reference Substance

Synonym(s):

4′,5,7-Trihydroxyflavone 7-rutinoside, Apigenin 7-rutinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O14
CAS Number:
Molecular Weight:
578.52
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

InChI

1S/C27H30O14/c1-10-20(31)22(33)24(35)26(38-10)37-9-18-21(32)23(34)25(36)27(41-18)39-13-6-14(29)19-15(30)8-16(40-17(19)7-13)11-2-4-12(28)5-3-11/h2-8,10,18,20-29,31-36H,9H2,1H3/t10-,18+,20-,21+,22+,23-,24+,25+,26+,27+/m0/s1

InChI key

FKIYLTVJPDLUDL-SLNHTJRHSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Isorhoifolin antioxidant properties research: A comprehensive study on the antidiabetic activities of Parquetina nigrescens identified Isorhoifolin as a key antidiabetic agent, underscoring its potent antioxidant properties in mitigating oxidative stress linked to diabetes (Ayoola et al., 2024).

  • Isorhoifolin flavonoid compound studies: The review of Mentha pulegium highlighted Isorhoifolin as a significant natural flavonoid, which exhibits diverse medicinal properties including anti-inflammatory and antioxidant activities, relevant for pharmacological innovations (Amtaghri et al., 2023).

  • Isorhoifolin applications in biochemistry: Investigation into the antidiabetic and cytotoxic activities of Gmelina philippensis emphasized Isorhoifolin′s role in biochemistry, showcasing its impact on cellular pathways through molecular docking studies and DFT calculations (Sayed et al., 2023).

  • Isorhoifolin effects on cellular pathways: Research on the biosynthesis of flavone rutinosides in Chrysanthemum species described the enzymatic roles that parallel compounds like Isorhoifolin play in plant biochemistry, affecting cellular pathways involved in plant defense and pigment production (Wu et al., 2022).

  • Isorhoifolin role in pharmacognosy research: A study on the major metabolites involved in the floral differentiation of Erythronium japonicum provided insights into Isorhoifolin′s effects at the molecular level, enhancing understanding of its pharmacognostic applications (Wang et al., 2022).

Other Notes

This compound is commonly found in plants of the genus: achillea mentha

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service