Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHL84266

Tubeimoside III

phyproof® Reference Substance

Synonym(s):

Tubeimoside C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C64H100O31
CAS Number:
Molecular Weight:
1365.46
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

CC1(C)CC[C@]2(C(O[C@@]3([H])[C@@](O[C@]4([H])O[C@@H](C)[C@]([H])(OC(C[C@@](O)(C)CC(OC5)=O)=O)[C@@H](O[C@@]6([H])[C@H](O)[C@@H](O)[C@H](O)CO6)[C@H]4O)([H])[C@@H](O)[C@@H](O)CO3)=O)[C@H](O)C[C@@]7(C)[C@]8(C)CC[C@@]9([H])[C@@](CO)(C)[C@@H](O[C@]%10([H])O[C@H

InChI

1S/C64H100O31/c1-25-47-48(91-52-44(79)38(73)29(68)21-85-52)46(81)54(87-25)92-49-39(74)30(69)22-86-55(49)95-57(82)64-14-13-58(2,3)15-27(64)26-9-10-34-60(5)16-28(67)51(61(6,24-66)33(60)11-12-62(34,7)63(26,8)17-35(64)70)94-56-50(43(78)40(75)31(20-65)88-56)93-53-45(80)42(77)41(76)32(89-53)23-84-36(71)18-59(4,83)19-37(72)90-47/h9,25,27-35,38-56,65-70,73-81,83H,10-24H2,1-8H3

InChI key

MTICHQXHYUJVDV-UHFFFAOYSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service