Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL84211

5,7-Dimethoxyflavone

phyproof® Reference Substance

Synonym(s):

5,7-Dimethoxy-2-phenyl-4H-1-benzopyran-4-one, Chrysin dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O4
CAS Number:
Molecular Weight:
282.29
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

SMILES string

O1c2c(c(cc(c2)OC)OC)C(=O)C=C1c3ccccc3

InChI

1S/C17H14O4/c1-19-12-8-15(20-2)17-13(18)10-14(21-16(17)9-12)11-6-4-3-5-7-11/h3-10H,1-2H3

InChI key

JRFZSUMZAUHNSL-UHFFFAOYSA-N

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wonhyoung Park et al.
Phytotherapy research : PTR, 34(9), 2275-2286 (2020-03-20)
Endometriosis is a reproductive disorder characterized by the dislocation of endometrial tissues. Approximately 5-20% of women at their reproductive age are diagnosed with endometriosis, which causes chronic pain and infertility. Here, we demonstrated that the bioactive flavonoid, 5,7-dimethoxyflavone (DMF), exhibited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service