Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHL83351

α-Humulene

phyproof® Reference Substance

Synonym(s):

α-Caryophyllene, trans,trans,trans-2,6,6,9-Tetramethyl-1,4,8-cycloundecatriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein:
3240075
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (GC)

form

liquid

manufacturer/tradename

PhytoLab

refractive index

n20/D 1.503 (lit.)

bp

166-168 °C (lit.)

density

0.889 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

C\C1=C/CC(C)(C)\C=C\C\C(C)=C\CC1

InChI

1S/C15H24/c1-13-7-5-8-14(2)10-12-15(3,4)11-6-9-13/h6-7,10-11H,5,8-9,12H2,1-4H3/b11-6+,13-7+,14-10+

InChI key

FAMPSKZZVDUYOS-HRGUGZIWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Anti-inflammatory activity: α-Humulene exhibits significant anti-inflammatory properties, making it a promising natural alternative for treating chronic inflammation and related disorders (Mahmoud et al., 2024).

  • Cancer research applications: The compound is under investigation for its potential to inhibit cancer cell growth, particularly through its impact on cell signaling pathways, which could lead to new cancer therapies (McGlinchey, 2024).

  • Pharmacological properties: α-Humulene is studied for its broad pharmacological effects, including its role in modulating immune responses and potential use in immune-related diseases (Cvetanović Kljakić et al., 2024).

  • Essential oil component: As a major component in various essential oils, α-Humulene contributes to their therapeutic benefits, such as antimicrobial and antioxidant activities, enhancing the value of these oils in natural medicine (Caruso et al., 2024).

  • Research on natural compound bioactivity: Ongoing studies focus on understanding the detailed mechanisms of α-Humulene′s action in cellular systems, aiming to harness its benefits in more targeted therapeutic applications (Li et al., 2024).


Other Notes

This compound is commonly found in plants of the genus: angelica salvia

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service