Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHL80067

Rebaudioside A

phyproof® Reference Substance

Synonym(s):

(4α)-13-[(2-O-β-D-glucopyranosyl-3-O-β-D­glucopyranosyl-β-D-glucopyranosyl)-oxy]kaur-6-en-8-oic acid β-D­glucopyranosyl ester, Glycoside A3, Glycoside X, Stevioside a3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H70O23
CAS Number:
Molecular Weight:
967.01
Beilstein:
6470556
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

C[C@@]12CCC[C@](C)([C@H]1CC[C@]34CC(=C)C(CC[C@@H]23)(C4)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)[C@H]5O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)C(=O)O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O

InChI

1S/C44H70O23/c1-17-11-43-9-5-22-41(2,7-4-8-42(22,3)40(59)66-38-33(58)30(55)26(51)20(14-47)62-38)23(43)6-10-44(17,16-43)67-39-35(65-37-32(57)29(54)25(50)19(13-46)61-37)34(27(52)21(15-48)63-39)64-36-31(56)28(53)24(49)18(12-45)60-36/h18-39,45-58H,1,4-16H2,2-3H3/t18-,19-,20-,21-,22+,23+,24-,25-,26-,27-,28+,29+,30+,31-,32-,33-,34+,35-,36+,37+,38+,39+,41-,42-,43-,44+/m1/s1

InChI key

HELXLJCILKEWJH-NCGAPWICSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Componentof stevia.

Application


  • Obesity managment: This study demonstrates the potential of Rebaudioside A derivatives like Rebaudioside D in reducing adiposity and liver fat accumulation, suggesting its utility in obesity management and metabolic syndrome research (Morissette et al., 2024).

  • Enzyme stability: The research includes the use of Rebaudioside A in enhancing enzyme stability, crucial for industrial applications such as sweetener production and pharmaceutical synthesis (Go et al., 2023).

  • Sugar-free products: The study investigates consumer perceptions of sugar-free products using Rebaudioside A, highlighting its importance in food science and consumer health (Williams et al., 2023).

  • Biotechnological applications: The study focuses on optimizing conditions for Rebaudioside A production in Stevia cultures, important for scaling up production in b (Ptak et al., 2023).

  • Sweetness and good taste: The study explores the genetic basis for Rebaudioside A biosynthesis, paving the way for genetic engineering approaches to enhance its production in Stevia plants (Li et al., 2023).

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service