Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

KRSU01

Sulfur in Odourless Kerosene

0.0005% (5 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material (UKAS ISO 17025 & ISO 17034 certified)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

concentration

0.0005% (5 ppm)

application(s)

petroleum

format

matrix material

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

168.8 °F

Flash Point(C)

76 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service