Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GARD02

Supelco

Gardner Colour Reference Standard

ASTM D 1544, D 6166 (ASTM colour 2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

according to ASTM® D 1544
according to ASTM® D 6166

shelf life

shelf life is based on the label and COFA in the package not on our website.

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Gardner colour reference standard is a certified reference material (CRM) certified to international standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation. Please note that the value stated on this page is nominal. All certified values for the current lots of all Gardner colour reference standards can be found on the Paragon Scientific webpage.

Application

Colour reference standard Gardner 2 is used to standardize color measuring instruments and validate test data. Its application is in process industries that employ colorimeters to assess the colour control of products such as oils, fuels, chemicals, pharmaceuticals, paints and coatings.

Features and Benefits

  • Produced and qualified according to ISO/IEC 17025 and ISO 17034 under UKAS accreditation
  • Colour values corresponding to ASTM D 1544/D 6166 colour range
  • Delivered with a certificate of calibration and MSDS
  • Available in 500-mL tamper-evident bottles
  • Full traceability to international standards with maximum data accuracy
  • 18-months shelf life

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service