Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G006163

HPLC Analysis of Exenatide Acetate on Proteomix® SCX-NP5

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Proteomix SCX NP5, 25 cm x 4.6mm I.D., 5 μm particles (Z777161)
mobile phase: (A) 10 mM monosodium phosphate:acetonitrile (56:44, v/v), adjusted to pH to 2.0 with phosphoric acid; ((B) 0.5 M NaCl/10 mM monosodium phosphate:acetonitrile (56:44, v/v), adjusted to pH to 2.0 with phosphoric acid)
gradient: 0 to 65% B in 35 min; held for 5 min; to 0% B in 10 min
flow rate: 1.0 mL/min
pressure: 1653 psi (114 bar)
column temp.: 40 °C
detector: UV, 235 nm
injection: 20 μL
sample: 0.325 mg/mL exenatide acetate in mobile phase A

suitability

application for HPLC

application(s)

life science and biopharma
pharmaceutical (small molecule)

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service