Skip to Content
MilliporeSigma
All Photos(1)

Documents

G006151

UHPLC/MS Analysis of Antineoplastic and Antidepressant Drugs in Plasma on Titan C18 after SPE using HybridSPE®-PLus

application for HPLC, application for SPE

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable
solid phase extraction (SPE): suitable

test parameters

sample/matrix: rat plasma
SPE well plate: HybridSPE-PLus 96-Well Plate, 50 mg/2 mL per well (575659-U)
sample preparation: final analyte concentration 50 ng/mL
sample addition: 400 μL of 1% formic acid acetonitile:spiked plasma (3:1) per well
column: Titan C18, 5 cm x 2.1 mm I.D., 1.9 μm particles (577122-U)
mobile phase: [A] 5 mM ammonium formate; [B] 5 mM ammonium formate in 90:10 (v:v) acetonitrile:water
gradient: 35 to 90% B in 1.5 min, held at 90% B for 0.4 min
flow rate: 0.4 mL/min
pressure: 3582 psi (247 bar)
column temp.: 35 °C
detector: TOF/MS
injection: 1 μL

suitability

application for HPLC
application for SPE

application(s)

pharmaceutical (small molecule)

Analysis Note

HybridSPE-Phospholipid plates removed plasma-derived proteins and phospholipids to maximize LC/MS sensitivity. Titan C18 U/HPLC columns provided rapid resolution. The highest grade LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.

Legal Information

HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Fluoxetine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    Tamoxifen, ≥99%

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service