Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G006141

HPLC Analysis of MAb on Proteomix® SCX-NP5 Comparing Different Buffer Systems and pH Gradients

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Proteomix SCX-NP5, 25 cm x 4.6 mm I.D., 5 μm particles (Z777161)
mobile phase: Mobile phase A: (A) 20 mM MES, pH 6.0; (B) A + 1 M NaCl, pH 6.0Mobile phase B: (A) 2.4 mM Tris, 1.5 mM imidazole, 11.6 mM piperazine, pH 6.0; (B) A, pH 10.5
gradient: Gradient A: 7% B held for 2 min; to 14% B in 20 min, Gradient B: 0 to 30% B in 2 min; to 55% B in 30 min
flow rate: 0.8 mL/min
detector: UV, 280 nm
injection: 5 μL
sample: 10 mg/mL

suitability

application for HPLC

application(s)

life science and biopharma
pharmaceutical (small molecule)

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service