Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G006136

HPLC Analysis of Proteins on Proteomix® SCX-NP1.7, Demonstration of Lot-to-Lot Reproducibility

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Proteomix SCX-NP1.7, 5 cm x 4.6 mm I.D., 1.7 μm particles (Z777146)
mobile phase: (A) 20 mM sodium phosphate buffer, pH 6.0, (B) A + 1.0 M NaCl
gradient: 0 - 75% B in 25 min, 15 min prewash
flow rate: 0.75 mL/min
column temp.: ambient
detector: UV, 214 nm
injection: 5 μL
sample: 1 mg/mL each compound

suitability

application for HPLC

application(s)

life science and biopharma
pharmaceutical (small molecule)

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Analyte

Description

    Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
    Cytochrome c from equine heart, ≥95% (SDS-PAGE)
    Lysozyme from chicken egg white, powder or granules, ≥90 %, ≥39,000 units/mg protein

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service