Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G006133

HPLC Analysis of HSA Fusion Protein on Proteomix® SAX-NP5

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

pressure: 2016 psi (139 bar)
column temp.: 30 °C
column: Proteomix SAX-NP5 25 cm x 4.6mm I.D., 5 μm particles (Z777229)
detector: UV, 280 nm
flow rate: 0.5 mL/min
mobile phase: Piperazine, Triethanolamine, bis-tris propane and N-methylpiperazine, 20 mM each,(A) pH 9.7; (B) pH 3.0 ( adjust by HCl )
sample: HSA fusion peptide 5 mg/mL, 10 μL (MW 75 kD, pI 5.0, HSA conjugated peptide in diabetes treatment)

suitability

application for HPLC

application(s)

life science and biopharma
pharmaceutical (small molecule)

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service