Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G006126

Protein Molecular Weight Calibration Curve on Zenix® Size Exclusion Columns, Effect of Pore Size

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Zenix SEC-150, 30 cm x 7.8 mm I.D., 3 μm, 150 Å particles (Z777018)
column: Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 μm, 300 Å particles (Z777033)
mobile phase: 150 mM sodium phosphate, pH 7
flow rate: 1 mL/min
column temp.: ambient
detector: UV, 214 nm
injection: 10 μL
sample: each 1 mg/mL except uracil, 2.5 mg/mL

suitability

application for HPLC

application(s)

life science and biopharma

Legal Information

Zenix is a registered trademark of Sepax Technologies

Analyte

Description

    Thyroglobulin from bovine thyroid, powder, ≥90% (agarose gel electrophoresis)
    γ-Globulins from human blood, ≥99% (electrophoresis)
    Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
    Ovalbumin (257-264) chicken, ≥97% (HPLC)
    Myoglobin from equine heart, ≥90% (SDS-PAGE), essentially salt-free, lyophilized powder
    Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
    Vanadium oxides tetroxide
    Uracil, ≥99.0% (T)
    See All (8)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service