Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G005596

HPLC Analysis of Polyphenols in Nero d′Avola Red Wine on Discovery® HS C18 (UV 354 nm)

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

sample/matrix: Nero d’Avola wine sample filtered prior injection through a 0.45 μm nylon membrane
column: Discovery HS C18, 15 cm x 2.1 mm I.D., 3 μm particles (569255-U)
mobile phase: [A] water, pH 3 with fomic acid; [B] Acetonitrile, pH 3 with fomic acid
flow rate: 0.2 mL/min
column temp.: 30 °C
detector: UV, 354 nm
injection: 5 μL

suitability

application for HPLC

application(s)

food and beverages

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Caftaric acid, analytical standard
    Rutin hydrate, ≥94% (HPLC), powder
    Isoquercitrin, primary reference standard

analytical column

standard


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service