Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G005595

HPLC Analysis of Polyphenols in Nero d′Avola Red Wine on Discovery® HS C18 (UV 280 nm)

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

sample/matrix: Nero d’Avola wine sample filtered prior injection through a 0.45 μm nylon membrane
column: Discovery HS C18, 15 cm x 2.1 mm I.D., 3 μm particles (569255-U)
mobile phase: [A] water, pH 3 with formic acid; [B] acetonitrile, pH 3 with formic acid
flow rate: 0.2 mL/min
column temp.: 30 °C
detector: UV, 280 nm
injection: 5 μL

suitability

application for HPLC

application(s)

food and beverages

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
    Caftaric acid, analytical standard
    (+)-Catechin, analytical standard
    Procyanidin B2, analytical standard
    Caffeic acid, ≥98.0% (HPLC)
    Ethyl gallate, antioxidant, ≥96.0% (HPLC)
    Ethyl gallate, antioxidant, ≥96.0% (HPLC)
    Rutin hydrate, ≥94% (HPLC), powder
    Isoquercitrin, primary reference standard
    p-Coumaric acid, ≥98.0% (HPLC), purum
    See All (10)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service