Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM47342

Supelco

Acrolein-DNPH solution

certified reference material, 1000 μg/mL in acetonitrile (as the aldehyde), ampule of 1 mL

Synonym(s):

Acrolein-2,4-dinitrophenylhydrazone solution, Acrolein-2,4-DNPH solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N4O4
CAS Number:
Molecular Weight:
236.18
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in acetonitrile (as the aldehyde)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

-10 to -25°C

SMILES string

[N+](=O)([O-])c1c(ccc(c1)[N+](=O)[O-])N\N=C\C=C

InChI

1S/C9H8N4O4/c1-2-5-10-11-8-4-3-7(12(14)15)6-9(8)13(16)17/h2-6,11H,1H2/b10-5+

InChI key

JJPZHGIYUVFTGG-BJMVGYQFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service