Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM31676

Supelco

Clodinafop-propargyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Topik, 2-Propynyl (R)-2-[4-(5-chloro-3-fluoro-2-pyridinyloxy)phenoxy]propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13ClFNO4
CAS Number:
Molecular Weight:
349.74
Beilstein:
8857008
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H](Oc1ccc(Oc2ncc(Cl)cc2F)cc1)C(=O)OCC#C

InChI

1S/C17H13ClFNO4/c1-3-8-22-17(21)11(2)23-13-4-6-14(7-5-13)24-16-15(19)9-12(18)10-20-16/h1,4-7,9-11H,8H2,2H3/t11-/m1/s1

InChI key

JBDHZKLJNAIJNC-LLVKDONJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Application

Clodinafop-propargyl is a herbicide for annual grass weed control in a range of crops (cereals including wheat, triticale and rye). It has a low aqueous solubility, is not volatile and is not expected to leach to groundwater. Clodinafop-propargyl is approved for use as plant protection product under Reg. (EC) No 1107/2009 in the European Union. Maximum residue levels (MRLs) have been set for various products of plant and animal origin according to Reg. (EU) No 777/2013 for clodinafop and its S-isomers and their salts, expressed as clodinafop from 0.02 to 0.1 mg/kg. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service