Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C16H13Cl3N2OS
CAS Number:
Molecular Weight:
387.71
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
grade
pharmaceutical primary standard
agency
BP
API family
tioconazole
form
solid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
BP
application(s)
pharmaceutical
pharmaceutical small molecule
format
mixture
storage temp.
2-8°C
SMILES string
Clc1ccc(C(Cn2ccnc2)OCc3ccsc3Cl)c(Cl)c1
InChI
1S/C16H13Cl3N2OS/c17-12-1-2-13(14(18)7-12)15(8-21-5-4-20-10-21)22-9-11-3-6-23-16(11)19/h1-7,10,15H,8-9H2
InChI key
QXHHHPZILQDDPS-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Tioconazole impurity standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.
Also used in monographs such as:Tioconazole Nail Solution
Also used in monographs such as:
Biochem/physiol Actions
Tioconazole is an antifungal medication of the imidazole class used to treat infections caused by fungus and yeast.It is more active than fluconazole or voriconazole against Candida glabrata mutant strains. It acts by membrane disruption and loss of intracellular ATP.
Packaging
Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia
Other Notes
Sales restrictions may apply.
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


