Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

96892

Supelco

1,4-Butanediol diglycidyl ether

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O4
CAS Number:
Molecular Weight:
202.25
Beilstein:
115238
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

vapor pressure

~10 mmHg ( 20 °C)

Assay

≥97.0% (GC)

technique(s)

HPLC: suitable

refractive index

n20/D 1.453 (lit.)

bp

266 °C (lit.)

density

1.1 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

C(CCOCC1CO1)COCC2CO2

InChI

1S/C10H18O4/c1(3-11-5-9-7-13-9)2-4-12-6-10-8-14-10/h9-10H,1-8H2

InChI key

SHKUUQIDMUMQQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service