Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94110

Supelco

Lutein (Zeaxanthin free)

analytical standard

Synonym(s):

Xanthophyll, α-Carotene-3,3′-diol, β, ε-carotene, β, ε-carotene-3,3′-diol, Lutein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56O2
CAS Number:
Molecular Weight:
568.87
Beilstein:
2068547
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

form

powder

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C

SMILES string

CC(=C/C=C/C=C(C)/C=C/C=C(C)/C=C/C1=C(C)C[C@@H](O)CC1(C)C)\C=C\C=C(C)\C=C\[C@H]2C(C)=C[C@H](O)CC2(C)C

InChI

1S/C40H56O2/c1-29(17-13-19-31(3)21-23-37-33(5)25-35(41)27-39(37,7)8)15-11-12-16-30(2)18-14-20-32(4)22-24-38-34(6)26-36(42)28-40(38,9)10/h11-25,35-37,41-42H,26-28H2,1-10H3/b12-11+,17-13+,18-14+,23-21+,24-22+,29-15+,30-16+,31-19+,32-20+/t35-,36+,37-/m0/s1

InChI key

KBPHJBAIARWVSC-RGZFRNHPSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

<ul>
<li><strong>Lutein in cellular antioxidant research:</strong> A study conducted by Lakshminarayana et al. investigates the oxidative derivatives of lutein in cellular environments using HeLa cells. The research focuses on lutein′s role as an antioxidant and its cytotoxic effects when exposed to oxidative stress, crucial for understanding its therapeutic potential in disease contexts where oxidative stress plays a role (Chemico-biological Interactions, 2013).</li>
<li><strong>Lutein in photosystem II research:</strong> Lutein binds violaxanthin, neoxanthin or zeaxanthin and even in the absence of zeaxanthin, plays a crucial role in the energy transfer processes within photosystems (European Journal of Biochemistry, 2001).</li>
</ul>

Biochem/physiol Actions

Dietary carotenoid with no vitamin A potency. Increases macular pigment concentration in the eye and may improve visual function.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service