Skip to Content
MilliporeSigma
All Photos(1)

Documents

91469

Supelco

Pyrazosulfuron-ethyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Ethyl 5-[(4,6-dimethoxy-2-pyrimidinyl)aminocarbonylaminosulfonyl]-1-methyl-1H-pyrazole-4-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N6O7S
CAS Number:
Molecular Weight:
414.39
Beilstein:
8442639
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

Assay

≥97.5%

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)c1cnn(C)c1S(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2

InChI

1S/C14H18N6O7S/c1-5-27-12(21)8-7-15-20(2)11(8)28(23,24)19-14(22)18-13-16-9(25-3)6-10(17-13)26-4/h6-7H,5H2,1-4H3,(H2,16,17,18,19,22)

InChI key

BGNQYGRXEXDAIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Application

Environmental
Pyrazosulfuron-ethyl is a herbicide mainly used to control broad-leaved weeds, grasses and sedges in rice. Pyrazosulfuron-ethyl is not listed in the European Union (EU) pesticide database. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service