Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91451

Supelco

Iprovalicarb

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Nα-(Isopropoxycarbonyl)-N-[1-(p-tolyl)ethyl]-L-valinamide, Isopropyl (S)-2-methyl-1-[1-(p-tolyl)ethylcarbamoyl]propylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28N2O3
CAS Number:
Molecular Weight:
320.43
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

SMILES string

CC(C1=CC=C(C=C1)C)NC([C@H](C(C)C)NC(OC(C)C)=O)=O

InChI

1S/C18H28N2O3/c1-11(2)16(20-18(22)23-12(3)4)17(21)19-14(6)15-9-7-13(5)8-10-15/h7-12,14,16H,1-6H3,(H,19,21)(H,20,22)/t14?,16-/m1/s1

InChI key

NWUWYYSKZYIQAE-BZSJEYESSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Iprovalicarb is a systematic fungicide with protective, eradicative, and curative activity against many oomycete pathogens. It is a mixture of (SS)-and (SR)-diastereoisomers that belong to the chemical class of amino acid amidocarbamate. Iprovalicarb is widely used for the control of potato late blight and downy mildew in many crops and shows excellent fungicidal activity against Plasmopara viticola, Peronospora vicia, Phytophthora sp., Alternaria sp. in grapes, potatoes, tomatoes, tobacco, and vegetables. The fungicide acts on the cell wall interrupting cellulose biosynthesis in pathogens.

According to Regulation (EC) 1107/2009, repealing Council Directive 91/414/EEC, iprovalicarb is approved for use in European Union (EU). The maximum residue level (MRL) following Reg. (EU) 777/2013 is set from 0.01 mg/kg to 2 mg/kg for products of plant and animal origin. ADI and AOEL of 0.015 mg/kg bw/day is established for iprovalicarb following Reg. (EU) 2016/147.

Iprovalicarb has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The iprovalicarb CRM may also find following uses:
  • To investigate the level of iprovalicarb resistance in Phytophthora infestans and the potential risk of resistance to iprovalicarb evolving in the future
  • Development of an enzyme-linked immunosorbent assay (ELISA) for the detection of iprovalicarb residues in agricultural and environmental samples
  • To analyze iprovalicarb residues in cabbage samples by high-performance liquid chromatography
  • To investigate the fate of indoxacarb, iprovalicarb, and boscalid residue in grapes and wine, after field treatment and during the winemaking process, by gas chromatography-ion trap mass spectrometry (GC-ITMS) analysis
  • To determine the baseline sensitivity of P. capsici to iprovalicarb by measuring mycelial growth
  • To develop and validate a method for the estimation of iprovalicarb residue in tomato fruit and grape berries and to study the persistence behavior of iprovalicarb in the selected food crops

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service