Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91011

Supelco

N-Phenyl-p-phenylenediamine

analytical standard

Synonym(s):

4-Aminodiphenylamine, Variamine Blue RT Base

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHC6H4NH2
CAS Number:
Molecular Weight:
184.24
Beilstein:
908935
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

68-72 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccc(Nc2ccccc2)cc1

InChI

1S/C12H12N2/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9,14H,13H2

InChI key

ATGUVEKSASEFFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

390.2 °F

Flash Point(C)

199 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service