Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

842197

Adapter for HPLC Columns, 1/4-28 to 10-32

PEEK, pkg of 1 ea

Synonym(s):

TSKgel® Adapter, PEEK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product name

Adapter, 1/4-28 to 10-32, PEEK, PEEK, pkg of 1 ea

material

PEEK

product line

TSKgel®

packaging

pkg of 1 ea

manufacturer/tradename

Tosoh 42197

technique(s)

HPLC: suitable

fitting

for 1/4-28 A
for 10-32 B

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service