Skip to Content

821970

TSKgel® NH2-100 (3 µm) HPLC Columns

L x I.D. 15 cm × 4.6 mm, amine phase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501
NACRES:
NB.21
L × i.d.:
15 cm × 4.6 mm
Particle size:
3 μm
Matrix active group:
NH2, amine phase
Pore size:
100 Å
Matrix:
silica particle platform
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

TSKgel® NH2-100 HPLC Column, phase amine, L × I.D. 15 cm × 4.6 mm, 3 μm particle size

material

stainless steel hardware

agency

suitable for USP L8

product line

TSKgel®

mol wt

100 - 2000

manufacturer/tradename

Tosoh 21970

parameter

0-100% organic solvent, 1.0 mL/min standard flow rate, 10-50 °C temperature, 150 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

450 m2/g

matrix

silica particle platform

matrix active group

NH2, amine phase

particle size

3 μm

pore size

100 Å

operating pH

2.0-7.5

separation technique

anion exchange, hydrophilic interaction (HILIC), normal phase

General description

TSKgel NH2-100 amino columns expand the range of TSKgel columns for hydrophilic interaction chromatography (HILIC). Offering a different selectivity from the well known TSKgel Amide-80 columns, these novel amino-bonded phase columns stand out by providing much improved chemical stability, a prerequisite for achieving reproducible and reliable results. TSKgel NH2-100 columns are packed with 3 micron silica particles containing 100Å pores. A novel bonding strategy was adopted to improve chemical stability of the bonded phase. First the silica is reacted with a trimethylsilane endcapping reagent at a low stoichiometric ratio, before reacting residual and accessible silanol groups with trifunctional alkylaminosilane reagent. The resulting bonded phase provides a better safeguard against hydrolysis of the underlying silica. TSKgel NH2-100 columns are unique in that the bonded phase ligand not only, as expected, has a terminal primary amino group, but that the spacer also incorporates secondary as well as tertiary amino groups. Anionic compounds are retained on the column by ionic interaction. This allows for the use of gradients in salt concentration, in addition to gradient elutions with acetonitrile. Since the TSKgel NH2-100 columns have cationic sites, the columns can be used as mixed mode columns under some conditions. TSKgel NH2-100 columns are well suited for the analysis of all types of hydrophilic compounds, including carbohydrates and peptides. Due to a high ligand density and large surface area, these columns show stronger retention of polar compounds than TSKgel Amide-80 columns.

Physical form

Shipped in 85% acetonitrile - 15% water.

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation


Still not finding the right product?

Explore all of our products under TSKgel® NH2-100 HPLC Column




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library