Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

821862

TSKgel® Amide-80 (3 µm) HPLC Columns

L x I.D. 1 cm × 2 mm, carbamoyl phase

Synonym(s):

TSKgel® Amide-80 Guard Cartridge, 1 cm x 2 mm ID, 3 μm, 3pk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® Amide-80 Guard Cartridge, phase carbamoyl, L × I.D. 1 cm × 2 mm, 3 μm particle size, pkg of 3 ea

material

stainless steel

Agency

suitable for USP L68

product line

TSKgel®

packaging

pkg of 3 ea

manufacturer/tradename

Tosoh 21862

parameter

0-100% organic solvent
10-50 °C temperature

technique(s)

HPLC: suitable

L × I.D.

1 cm × 2 mm

matrix

silica particle platform

matrix active group

carbamoyl phase

particle size

3 μm

pore size

100 Å

operating pH

2.0-7.5

separation technique

hydrophilic interaction (HILIC)

Looking for similar products? Visit Product Comparison Guide

General description

Guard Cartridge for 2mmID TSKgel Amide-80 columns, 3μm, 3pk
TSKgel Guard cartridges require a holder, which is sold separately. For 2 mm cartridges, the holder is Part No. 819308. For the 3.2 mm cartridges used to protect 4.6 mm columns, the holder is Part No. 819018.

Physical form

Shipped in 85% acetonitrile - 15% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

required but not provided

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service