Skip to Content
MilliporeSigma
All Photos(1)

Documents

82030-U

Hamilton® PRP-1 (5 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, PEEK hardware

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Hamilton PRP-1 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm, PEEK hardware

material

PEEK column

Agency

suitable for USP L21

manufacturer/tradename

Hamilton

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

PS-DVB particle platform
polymeric

matrix active group

polymer

particle size

5 μm

pore size

100 Å

operating pH

1-14

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

The Hamilton PRP-1, polymeric reversed phase columns are Ideal for high pH (pH 8-13) operation or with analytes that give poor chromatogrpahy on silica-based columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service