Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

813062

TSKgel® SP-5PW (10 µm) HPLC Columns

L x I.D. 5 cm × 5 mm, sulfonic acid phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
L × I.D.:
5 cm × 5 mm
particle size:
10 μm
matrix active group:
sulfonic acid phase
matrix:
hydroxylated methacrylic polymer

Product Name

TSKgel® SP-5PW HPLC Column, phase sulfonic acid, L × I.D. 5 cm × 5 mm, 10 μm particle size

material

glass

product line

TSKgel®

feature

counter ion Na+

manufacturer/tradename

Tosoh 13062

parameter

≤1 M salt concentration
≤20% organic solvent
1.0 mL/min flow rate
10-45 °C temperature
150 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

5 cm × 5 mm

matrix

hydroxylated methacrylic polymer

matrix active group

sulfonic acid phase

particle size

10 μm

operating pH

2-12

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel® SP-5PW columns are packed with porous hydrophilic polymer beads which are surface modified with a strong cation exchanger. They are used for the separation and analysis of proteins, peptides, and other biologically active molecules. TSKgel SP-5PW columns have the same backbone chemistry and selectivity as the bulk process scale resins Toyopearl SP-650 and TSKgel SP-5PW (20) and (30).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service